XL765 ≥95%

Supplier: ADIPOGEN CORP MS

SYN-1137-M001 SYN-1137-M005 SYN-1137-M010 SYN-1137-M050
102992-296EA 186.92 USD
102992-296 102992-298 102992-300 102992-302
XL765 ≥95%
XL765

XL765 is active against class I PI3K (IC(50) = 39, 113, 9 and 43nM for p110alpha, beta, gamma and delta, respectively). XL765 also inhibits DNA-PK (IC(50) = 150nM) and mTOR (IC(50) = 157nM)

Formula: C₁₃H₁₄N₆O
Storage Temperature: Freezer
MDL Number: MFCD20276656
CAS Number: 1349796-36-6

Order Now

Specification Test Results

Purity >95%
Appearance/Color Solid.
Solubility Soluble in DMSO. Slightly soluble (<1mg/ml) in ethanol.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR