NVP-BSK805 dihydrochloride ≥95%
Supplier: ADIPOGEN CORP MS
Synonyms:
4-(2,6-difluoro-4-(3-(1-(pip4-(2,6-difluoro-4-(3-(1-(piperidin-4-yl)-1H-pyrazol-4-yl)quinoxalin-5-yl)benzyl)morpholine dihydrochloride
SYN-1136-M001
SYN-1136-M005
SYN-1136-M010
SYN-1136-M050
102992-288EA
246.46
USD
102992-288
102992-290
102992-292
102992-294
NVP-BSK805 dihydrochloride ≥95%
NVP-BSK805 dihydrochloride
NVP-BSK805 is a potent and selective quinoxaline JAK2 inhibitor with IC(50) values of 0.48, 0.56 and 0.58nM for JAK2 JH1, FL JAK (V617F) and FL JAK2 wt, respectively.
Formula:
C₂₇H₂₈F₂N₆O.2HCl MW: 563.5 g/mol Storage Temperature: Freezer |
CAS Number:
1942919-79-0 |
Specification Test Results
Purity | >95% |
Appearance/Color | Solid. |
Solubility | Soluble in DMSO or ethanol. |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...