Brivanib alaninate ≥95%

Supplier: ADIPOGEN CORP MS

Synonyms: BMS-582664, (R)-(S)-1-((4-((4-Fluoro-2-methyl-1H-indol-5-yl)oxy)-5-methylpyrrolo[2,1-f][1,2,4]triazin-6-yl)oxy)propan-2-yl 2-aminopropanoate, Brivanib (alaninate)

SYN-1135-M001 SYN-1135-M005 SYN-1135-M010 SYN-1135-M050
102992-280EA 217.38 USD
102992-280 102992-282 102992-284 102992-286
Brivanib alaninate ≥95%
Brivanib alaninate

Brivanib inhibits VEGFR-2 with an IC(50) of 25nM and a Ki of 26nM, VEGFR-1 with an IC(50) of 380nM and VEGFR-3 with an IC(50) of 10nM. It also shows selectivity for FGFR-1 with an IC(50) of 148nM, FGFR-2 with an IC(50) of 25nM and FGFR-3 with an IC(50) of 68nM.

Formula: C₂₂H₂₄FN₅O₄
MW: 441.5 g/mol
Storage Temperature: Freezer
MDL Number: MFCD22577162
CAS Number: 649735-63-7

Order Now

Specification Test Results

Purity 95%
Appearance/Color Solid.
Solubility Soluble in DMSO or ethanol.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR