JNJ-38158471 ≥95%
Supplier: ADIPOGEN CORP MS
Synonyms:
JNJ38158471, 1-(4-((6-Amino-5-((methoxyimino)methyl)pyrimidin-4-yl)oxy)-2-chlorophenyl)-3-ethylurea
SYN-1133-M001
SYN-1133-M005
SYN-1133-M010
102992-266EA
283.85
USD
102992-266
102992-268
102992-270
JNJ-38158471 ≥95%
JNJ-38158471
JNJ-38158471 is a well tolerated, orally available, highly selective VEGFR-2 inhibitor with an IC(50) of 40nM. It also inhibits closely related tyrosine kinases such as Ret (IC(50) of 180nM) and Kit (C50 of 500nM). It has no significant activity (
Formula:
C₁₅H₁₇ClN₆O₃ Storage Temperature: Freezer |
MDL Number:
MFCD22631501 CAS Number: 951151-97-6 |
Specification Test Results
Purity |
|
Appearance/Color | Solid. |
Solubility | Soluble in DMSO. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...