S6K-18 ≥95%

Supplier: ADIPOGEN CORP MS

Synonyms: S6K1-IN-1, 2-(3-(1H-Indazol-5-yl)ureido)-5-(tert-butyl)thiophene-3-carboxylic acid

SYN-1132-M001 SYN-1132-M005 SYN-1132-M010
102992-260EA 283.85 USD
102992-260 102992-262 102992-264
S6K-18 ≥95%
S6K-18

S6K-18 is a highly selective inhibitor of S6K1, with an IC(50) of 52nM.

Formula: C₁₇H₁₈N₄O₃S
MW: 358.4 g/mol
Storage Temperature: Freezer
MDL Number: MFCD22631500
CAS Number: 1265789-88-5

Order Now

Specification Test Results

Purity 95%
Appearance/Color Solid.
Solubility Soluble in DMSO.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR