Bayer-18 ≥95%
Supplier: ADIPOGEN CORP MS
|
Warning
|
SYN-1130-M001
SYN-1130-M005
SYN-1130-M010
102992-248EA
283.85
USD
102992-248
102992-250
102992-252
Bayer-18 ≥95%
Bayer-18
Bayer-18 is a selective TYK2 inhibitor, with an IC(50) of 18.7nM on TYK2 as measured by TYK2 HTRF assays. The compound shows 1000 fold selectivity against JAK2, CDK and KDR kinases in similar HTRF assays.
Formula:
C₁₉H₂₇FN₆O₂ Storage Temperature: Freezer |
CAS Number:
1251752-12-1 |
Specification Test Results
Purity |
|
Appearance/Color | Solid. |
Solubility | Soluble in DMSO. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...