Foretinib ≥95%

Supplier: ADIPOGEN CORP MS

Synonyms: N-(3-Fluoro-4-((6-methoxy-7-(3-morpholinopropoxy)quinolin-4-yl)oxy)phenyl)-N-(4-fluorophenyl)cyclopropane-1,1-dicarboxamide, GSK-1363089

SYN-1129-M001 SYN-1129-M005 SYN-1129-M010 SYN-1129-M050
102992-240EA 156.46 USD
102992-240 102992-242 102992-244 102992-246
Foretinib ≥95%
Foretinib

Foretinib is a novel MET and VEGFR2 kinase inhibitor with IC(50) values of 0.4 and 0.8nM for MET and VEGFR2 respectively.

Formula: C₃₄H₃₄F₂N₄O₆
MW: 632.7 g/mol
Storage Temperature: Freezer
MDL Number: MFCD16038048
CAS Number: 849217-64-7

Order Now

Specification Test Results

Purity 95%
Appearance/Color Solid.
Solubility Soluble in DMSO.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR