GSK2126458 ≥95%
Supplier: ADIPOGEN CORP MS
|
Warning
|
Synonyms:
HYR-582, Omipalisib, 2,4-Difluoro-N-(2-methoxy-5-(4-(pyridazin-4-yl)quinolin-6-yl)pyridin-3-yl)benzenesulphonamide
GSK2126458 is an orally available selective inhibitor of the class I phosphoinositide 3-kinase (PI3K) enzymesand MTOR1/2 complexes. GSK2126458 can inhibit PI3K-alpha (p85alpha/p110alpha) with IC(50) of 0.04nM. GSK2126458 has Ki values in the picomolar range for each of the class I PI3K isoforms and MTOR1/2 complexes. GSK2126458 has potent in vitro and in vivo growth-inhibitory effects on cancer cells. In comparison with other clinical PI3K inhibitors, GSK2126458 is around 100-fold more potent than BEZ235 (6nM). GSK2126458 also a low picomolar inhibitor of the common activating mutants of p110R ( E542K, E545K, and H1047R ) with Ki of 0.008nM, 0.008nM and 0.009nM in human cancer. In mechanistic cellular assays, GSK2126458 caused a significant reduction in the levels of pAKT-S473 with remarkable potency. Consistent with its activity against both PI3K R and mTOR, GSK2126458 also inhibits phosphorylation of AKT-T308 and p70S6K at low nanomolar concentrations. GSK2126458 induces a G1 cell cycle arrest and inhibits cell proliferation in a large panel of cell lines, including T47D and BT474 breast cancer lines.
Formula:
C₂₅H₁₇F₂N₅O₃S MW: 505.5 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD16038929 CAS Number: 1086062-66-9 |
Specification Test Results
Purity |
|
Appearance/Color | Solid. |
Solubility |
Soluble in DMSO. Slightly soluble ( |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...