VX-745 ≥95%

Supplier: ADIPOGEN CORP MS
Warning

Synonyms: 5-(2,6-Dichlorophenyl)-2-((2,4-difluorophenyl)thio)-6H-pyrimido[1,6-b]pyridazin-6-one, Neflamapimod

SYN-1123-M001 SYN-1123-M005 SYN-1123-M010 SYN-1123-M050
102992-198EA 119.08 USD
102992-198 102992-200 102992-202 102992-204
VX-745 ≥95%
VX-745

VX745 is a selective inhibitor of p38alpha with an IC(50) of 10nM.

Formula: C₁₉H₉Cl₂F₂N₃OS
MW: 436.3 g/mol
Storage Temperature: Freezer
MDL Number: MFCD09834070
CAS Number: 209410-46-8

Order Now

Specification Test Results

Purity 95%
Appearance/Color Solid.
Solubility Soluble in DMSO or ethanol.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR