R406 free base ≥95%

Supplier: ADIPOGEN CORP MS
Warning

Synonyms: 6-[5-Fluoro-2-(3,4,5-trimethoxyphenylamino)pyrimidin-4-ylamino]-2,2-dimethyl-4H-pyrido[3,2-b][1,4]oxazin-3-one

SYN-1121-M001 SYN-1121-M005 SYN-1121-M010 SYN-1121-M050
102992-182EA 149.54 USD
102992-182 102992-184 102992-186 102992-188
R406 free base ≥95%
R406 free base

R406 is a strong Syk inhibitor with an IC(50) of 41nM. It inhibits Syk but not Lyn. It shows 5-fold less potency to Flt3.

Formula: C₂₂H₂₃FN₆O₅
MW: 470.45 g/mol
Storage Temperature: Freezer
MDL Number: MFCD09970820
CAS Number: 841290-80-0

Order Now

Specification Test Results

Purity >95%
Appearance/Color Solid.
Solubility Soluble in DMSO or ethanol.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR