Baricitinib phosphate salt ≥95%
Supplier: ADIPOGEN CORP MS
Baricitinib is a selective JAK1 and JAK2 inhibitor. IC(50) values are 5.9nm for JAK1, and 5.7nM for JAK2. In cell based assays Baricitinib/INCB-28050 demonstrated inhibition of intracellular signaling of multiple pro-inflammatory cytokines including IL-6 and IL-23 at concentrations of
Formula:
C₁₆H₁₇N₇O₂S·H₃PO₄ Storage Temperature: Freezer |
MDL Number:
MFCD25976707 CAS Number: 1187595-84-1 |
Specification Test Results
Purity |
|
Appearance/Color | Solid. |
Solubility | Soluble in DMSO or ethanol. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...