Ponatinib ≥95%
Supplier: ADIPOGEN CORP MS
SYN-1116-M001
SYN-1116-M005
SYN-1116-M010
SYN-1116-M050
102992-146EA
90
USD
102992-146
102992-148
102992-150
102992-152
Ponatinib ≥95%
Ponatinib
Ponatinib is a potent multi-target inhibitor of Abl, PDGFRalpha, VEGFR2, FGFR1 and Src with IC(50) values of 0.37, 1.1, 1.5, 2.2 and 5.4nM respectively.
Formula:
C₂₉H₂₇F₃N₆O Storage Temperature: Freezer |
MDL Number:
MFCD17215203 CAS Number: 943319-70-8 |
Specification Test Results
Purity |
|
Appearance/Color | Solid. |
Solubility | Soluble in DMSO or ethanol. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...