GNE-493 ≥95%

Supplier: ADIPOGEN CORP MS

Synonyms: 2-(2-(2-Aminopyrimidin-5-yl)-4-morpholinothieno[3,2-d]pyrimidin-6-yl)propan-2-ol

SYN-1115-M001 SYN-1115-M005 SYN-1115-M010
102992-140EA 283.85 USD
102992-140 102992-142 102992-144
GNE-493 ≥95%
GNE-493

GNE-493 is a potent, selective, and orally available pan-PI3-kinase and dual pan-PI3-kinase/mTOR inhibitor for the treatment of cancer.

Formula: C₁₇H₂₀N₆O₂S
MW: 372.4 g/mol
Storage Temperature: Freezer
MDL Number: MFCD18251526
CAS Number: 1033735-94-2

Order Now

Specification Test Results

Purity 95%
Appearance/Color Solid.
Solubility Soluble in DMSO.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR