Mubritinib ≥95%
Supplier: ADIPOGEN CORP MS
Synonyms:
(E)-4-[[4-[4-(1H-1,2,3-Triazol-1-yl)butyl]phenoxy]methyl]-2-[4-(trifluoromethyl)styryl]oxazole, TAK 165, (E)-4-((4-(4-(1H-1,2,3-Triazol-1-yl)butyl)phenoxy)methyl)-2-(4-(trifluoromethyl)styryl)oxazole
SYN-1112-M001
SYN-1112-M005
SYN-1112-M010
SYN-1112-M050
102992-118EA
74.77
USD
102992-118
102992-120
102992-122
102992-124
Mubritinib ≥95%
Mubritinib
Mubritinib is a potent, irreversible human epithelial growth factor receptor 2 (ErbB2/HER2) inhibitor (IC(50) of 6nM) which displays
Formula:
C₂₅H₂₃F₃N₄O₂ MW: 468.5 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD09954135 CAS Number: 366017-09-6 |
Specification Test Results
Purity |
|
Appearance/Color | Solid. |
Solubility | Soluble in DMSO or ethanol. |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...