CX-4945 ≥95%

Supplier: ADIPOGEN CORP MS
Warning

Synonyms: Silmitasertib, 5-((3-Chlorophenyl)amino)benzo[c][2,6]naphthyridine-8-carboxylic acid

SYN-1109-M001 SYN-1109-M005 SYN-1109-M010 SYN-1109-M050
102992-098EA 149.54 USD
102992-098 102992-100 102992-102 102992-104
CX-4945 ≥95%
CX-4945

CX-4945 is a first-in-class, orally active, potent and selective inhibitor of the protein kinase CK2, with an IC(50) of 1nM.

Formula: C₁₉H₁₂ClN₃O₂
MW: 349.8 g/mol
Storage Temperature: Freezer
MDL Number: MFCD13184796
CAS Number: 1009820-21-6

Order Now

Specification Test Results

Purity 95%
Appearance/Color Solid.
Solubility Soluble in DMSO or ethanol.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR