TG-89 ≥95%

Supplier: ADIPOGEN CORP MS

Synonyms: N-(tert-Butyl)-3-((5-methyl-2-((3-(morpholinomethyl)phenyl)amino)pyrimidin-4-yl)amino)benzenesulphonamide

SYN-1107-M001 SYN-1107-M005 SYN-1107-M010
102992-084EA 283.85 USD
102992-084 102992-086 102992-088
TG-89 ≥95%
TG-89

TG-89 is a JAK inhibitor. It has IC(50) values of 6, 25, 17 and 169nM against JAK2, FLT3, RET and JAK3 respectively.

Formula: C₂₆H₃₄N₆O₃S
MW: 510.7 g/mol
Storage Temperature: Freezer
CAS Number: 936091-56-4

Order Now

Specification Test Results

Purity 95%
Appearance/Color Solid.
Solubility Soluble in DMSO or ethanol.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR