TG-46 ≥95%
Supplier: ADIPOGEN CORP MS
Synonyms:
JAK-IN-35, N-(tert-Butyl)-3-((5-methyl-2-((4-(morpholinomethyl)phenyl)amino)pyrimidin-4-yl)amino)benzenesulphonamide
SYN-1106-M001
SYN-1106-M005
SYN-1106-M010
102992-078EA
283.85
USD
102992-078
102992-080
102992-082
TG-46 ≥95%
TG-46
TG-46 is an inhibitor of JAK2, FLT3, RET, JAK3 with IC(50) values of 6nM, 25nM, 17nM, 169nM respectively.
Formula:
C₂₆H₃₄N₆O₃S MW: 510.7 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD17019342 CAS Number: 936091-15-5 |
Specification Test Results
Purity |
|
Appearance/Color | Solid. |
Solubility | Soluble in DMSO or ethanol. |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...