TG100115 ≥95%

Supplier: ADIPOGEN CORP MS

Synonyms: TG100-115, 3,3'-(2,4-Diaminopteridine-6,7-diyl)diphenol

SYN-1105-M001 SYN-1105-M005 SYN-1105-M010 SYN-1105-M050
102992-070EA 149.54 USD
102992-070 102992-072 102992-074 102992-076
TG100115 ≥95%
TG100115

TG100115 inhibits PI3Kgamma and delta, with IC(50) values of 83 and 235nM, respectively.

Formula: C₁₈H₁₄N₆O₂
MW: 346.3 g/mol
Storage Temperature: Freezer
MDL Number: MFCD16038873
CAS Number: 677297-51-7

Order Now

Specification Test Results

Purity 95%
Appearance/Color Solid.
Solubility Soluble in DMSO. Slightly soluble (1mg/ml) in ethanol.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR