ZSTK474 ≥95%

Supplier: ADIPOGEN CORP MS
Warning

Synonyms: 4,4'-(6-(2-(Difluoromethyl)-1H-benzo[d]imidazol-1-yl)-1,3,5-triazine-2,4-diyl)dimorpholine

SYN-1099-M001 SYN-1099-M005 SYN-1099-M010 SYN-1099-M050
102992-032EA 74.77 USD
102992-032 102992-034 102992-036 102992-038
ZSTK474 ≥95%
ZSTK474

ZSTK474 Inhibits all four PI3K isoforms in an ATP-competitive manner. IC(50) is approximately 37nM.

Formula: C₁₉H₂₁F₂N₇O₂
MW: 417.4 g/mol
Storage Temperature: Freezer
MDL Number: MFCD08705334
CAS Number: 475110-96-4

Order Now

Specification Test Results

Purity 95%
Appearance/Color Solid.
Solubility Soluble in DMSO. Slightly soluble (1mg/ml) in ethanol.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR