Intedanib ≥95%
Supplier: ADIPOGEN CORP MS
Synonyms:
BIBF 1120, Methyl (Z)-3-(((4-(N-methyl-2-(4-methylpiperazin-1-yl)acetamido)phenyl)amino)(phenyl)methylene)-2-oxoindoline-6-carboxylate, Nintedanib, Vargatef
SYN-1091-M001
SYN-1091-M005
SYN-1091-M010
SYN-1091-M050
102991-972EA
119.08
USD
102991-972
102991-974
102991-976
102991-978
Intedanib ≥95%
Intedanib
Intedanib is an indolinone derivative that potently inhibits VEGFR1/2/3, FGFR1/2/3 and PDGFRalpha/beta with IC(50) values of 34, 13, 13, 69, 37, 108, 59 and 65nM.
Formula:
C₃₁H₃₃N₅O₄ MW: 539.6 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD11974012 CAS Number: 656247-17-5 |
Specification Test Results
Purity |
|
Appearance/Color | Solid. |
Solubility | Soluble in DMSO. |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...