TGX-221 ≥95%

Supplier: ADIPOGEN CORP MS
Warning

SYN-1089-M001 SYN-1089-M005 SYN-1089-M010 SYN-1089-M050
102991-956EA 96.92 USD
102991-956 102991-958 102991-960 102991-962
TGX-221 ≥95%
TGX-221

TGX221 is a potent, cell permeable ATP-competitive inhibitor of PI3K, with selectivity for the p110beta isoform (IC(50) of 50nM).

Formula: C₂₁H₂₄N₄O₂
Storage Temperature: Freezer
MDL Number: MFCD11113209
CAS Number: 663619-89-4

Order Now

Specification Test Results

Purity 95%
Appearance/Color Solid.
Solubility Soluble in DMSO. Slightly soluble (1mg/ml) in ethanol.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR