Tandutinib ≥95%

Supplier: ADIPOGEN CORP MS
Warning

Synonyms: N-(4-Isopropoxyphenyl)-4-(6-methoxy-7-(3-(piperidin-1-yl)propoxy)quinazolin-4-yl)piperazine-1-carboxamide, CT53518

SYN-1088-M001 SYN-1088-M005 SYN-1088-M010 SYN-1088-M050
102991-948EA 74.77 USD
102991-948 102991-950 102991-952 102991-954
Tandutinib ≥95%
Tandutinib

Tandutinib inhibits the auto-phosphorylation of FLT3 (FMS-Like Tyrosine kinase-3, IC(50) of 0.22µM), c-KIT and PDGF (platelet-derived growth factor), thereby inhibiting cellular proliferation and inducing apoptosis.

Formula: C₃₁H₄₂N₆O₄
MW: 562.7 g/mol
Storage Temperature: Freezer
MDL Number: MFCD09954147
CAS Number: 387867-13-2

Order Now

Specification Test Results

Purity 95%
Appearance/Color Solid.
Solubility Soluble in DMSO or ethanol.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR