TAK-715 ≥95%

Supplier: ADIPOGEN CORP MS
Warning

Synonyms: N-(4-(2-Ethyl-4-(m-tolyl)thiazol-5-yl)pyridin-2-yl)benzamide

SYN-1087-M001 SYN-1087-M005 SYN-1087-M010 SYN-1087-M050
102991-940EA 105.23 USD
102991-940 102991-942 102991-944 102991-946
TAK-715 ≥95%
TAK-715

TAK-715 is a potent specific inhibitor of the p38alpha MAP kinase with an IC(50) of 7.1nM. It is approximately 30-fold more selective for p38alpha over p38beta. It shows no inhibition of p38gamma/delta.

Formula: C₂₄H₂₁N₃OS
MW: 399.5 g/mol
Storage Temperature: Freezer
MDL Number: MFCD17012805
CAS Number: 303162-79-0

Order Now

Specification Test Results

Purity 95%
Appearance/Color Solid.
Solubility Soluble in DMSO or ethanol.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR