SB590885 ≥95%
Supplier: ADIPOGEN CORP MS
Synonyms:
5-(2-(4-(2-Dimethylaminoethoxy)phenyl)-5-(4-pyridinyl)-1H-imidazol-4-yl)-2,3-dihydro-1H-inden-1-one oxime, 5-(2-(4-(2-(Dimethylamino)ethoxy)phenyl)-5-(pyridin-4-yl)-1H-imidazol-4-yl)-2,3-dihydro-1H-inden-1-one oxime
SYN-1077-M001
SYN-1077-M005
SYN-1077-M010
SYN-1077-M050
102991-862EA
119.08
USD
102991-862
102991-864
102991-866
102991-868
SB590885 ≥95%
SB590885
SB-590885 is a novel, potent and selective small molecule inhibitor of B-Raf kinase, with a Ki of 0.16nM. It shows an 11-fold greater selectivity for B-Raf over c-Raf. It was used to address the dependence of MAPK signalling and tumor cell growth on B-Raf kinase activity.
Formula:
C₂₇H₂₇N₅O₂ Storage Temperature: Freezer |
MDL Number:
MFCD16038645 CAS Number: 405554-55-4 |
Specification Test Results
Purity | >95% |
Appearance/Color | Solid. |
Solubility | Soluble in DMSO or ethanol. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...