SB216763 ≥95%

Supplier: ADIPOGEN CORP MS
Warning

Synonyms: 3-(2,4-Dichlorophenyl)-4-(1-methyl-1H-indol-3-yl)-1H-pyrrole-2,5-dione, SB-216763

SYN-1075-M001 SYN-1075-M005 SYN-1075-M010 SYN-1075-M050
102991-848EA 119.08 USD
102991-848 102991-850 102991-852 102991-854
SB216763 ≥95%
SB216763

SB-216763 is a potent and selective ATP-competitive GSK-3 inhibitor with an IC(50) of 34.3nM for GSK-3alpha, and similar activity for GSK-3beta.

Formula: C₁₉H₁₂Cl₂N₂O₂
MW: 371.2 g/mol
Storage Temperature: Freezer
MDL Number: MFCD09753369
CAS Number: 280744-09-4

Order Now

Specification Test Results

Purity 95%
Appearance/Color Solid.
Solubility Soluble in DMSO or ethanol.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR