SB202190 hydrochloride ≥95%
Supplier: ADIPOGEN CORP MS
|
Warning
|
SYN-1073A-M001
SYN-1073A-M005
SYN-1073A-M010
SYN-1073A-M050
102991-832EA
74.77
USD
102991-832
102991-834
102991-836
102991-838
SB202190 hydrochloride ≥95%
SB202190 hydrochloride
SB202190 hydrochloride is a water soluble form of the potent p38 (SAPK2a) inhibitor SB202190.
Formula:
C₂₀H₁₄FN₃O·HCl Storage Temperature: Freezer |
CAS Number:
350228-36-3 |
Specification Test Results
Purity |
|
Appearance/Color | Solid. |
Solubility | Soluble in DMSO or ethanol. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...