GSK429286A ≥95%
Supplier: ADIPOGEN CORP MS
Synonyms:
N-(6-Fluoro-1H-indazol-5-yl)-2-methyl-6-oxo-4-(4-(trifluoromethyl)phenyl)-1,4,5,6-tetrahydropyridine-3-carboxamide, 4-[4-(Trifluoromethyl)phenyl]-N-(6-Fluoro-1H-indazol-5-yl)-2-methyl-6-oxo-1,4,5,6-tetrahydro-3-pyridinecarboxamide, GSK429286, RHO-15
SYN-1071-M001
SYN-1071-M005
SYN-1071-M010
SYN-1071-M050
102991-808EA
134.31
USD
102991-808
102991-810
102991-812
102991-814
GSK429286A ≥95%
GSK429286A
GSK429286A is a potent and selective inhibitor of Rho-associated kinase (ROCK1), with an IC(50) of 14nM. It is orally available and has been shown to dramatically reduce mean arterial pressure in spontaneously hypertensive rats.
Formula:
C₂₁H₁₆F₄N₄O₂ MW: 432.4 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD16618404 CAS Number: 864082-47-3 |
Specification Test Results
Purity |
|
Appearance/Color | Solid. |
Solubility | Soluble in DMSO. |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...