Purvalanol B ≥95%

Supplier: ADIPOGEN CORP MS
Warning

Synonyms: NG95, (R)-2-Chloro-4-((2-((1-hydroxy-3-methylbutan-2-yl)amino)-9-isopropyl-9H-purin-6-yl)amino)benzoic acid

SYN-1070-M001 SYN-1070-M005 SYN-1070-M010 SYN-1070-M050
102991-800EA 74.77 USD
102991-800 102991-802 102991-804 102991-806
Purvalanol B ≥95%
Purvalanol B

Purvalanol B is a cyclin-dependent kinase inhibitor with IC(50) values of 6, 6, 9, 10,000, and 6nM for cdc2/cyclin B, cdk2/cyclin A, cdk2/cyclin E, cdk4/cyclin D1 and cdk5-p35, respectively. Purvalanol B has also been shown to have anti-proliferative properties, mediated by p42/p44 MAPK.

Formula: C₂₀H₂₅ClN₆O₃
MW: 432.9 g/mol
Storage Temperature: Freezer
CAS Number: 212844-54-7

Order Now

Specification Test Results

Purity 95%
Appearance/Color Solid.
Solubility Soluble in DMSO or ethanol.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR