PIK-90 ≥95%
Supplier: ADIPOGEN CORP MS
Synonyms:
N-(7,8-Dimethoxy-2,3-dihydroimidazo[1,2-c]quinazolin-5-yl)nicotinamide
SYN-1068-M001
SYN-1068-M005
SYN-1068-M010
SYN-1068-M050
102991-784EA
74.77
USD
102991-784
102991-786
102991-788
102991-790
PIK-90 ≥95%
PIK-90
PIK-90 is a potent PI3K inhibitor with IC(50) values of 11, 18, and 58nM for p110alpha, gamma and delta isoforms respectively. It has an IC(50) of 350nM for the p110beta isoform. It also has low mTOR activity.
Formula:
C₁₈H₁₇N₅O₃ MW: 351.4 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD12910483 CAS Number: 677338-12-4 |
Specification Test Results
Purity |
|
Appearance/Color | Solid. |
Solubility |
Slightly soluble ( |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...