PIK-75 ≥95%
Supplier: ADIPOGEN CORP MS
|
Warning
|
Synonyms:
N'-((6-Bromoimidazo[1,2-a]pyridin-3-yl)methylene)-N,2-dimethyl-5-nitrobenzenesulphonohydrazide
SYN-1067-M001
SYN-1067-M005
SYN-1067-M010
SYN-1067-M050
102991-776EA
74.77
USD
102991-776
102991-778
102991-780
102991-782
PIK-75 ≥95%
PIK-75
PIK-75 is a PI3K inhibitor displaying selectivity for the p110alpha isoform (IC(50) of 6nM). PIK-75 blocks the phosphorylation of PKB induced by insulin.
Formula:
C₁₆H₁₄BrN₅O₄S MW: 452.3 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD12407411 CAS Number: 372196-67-3 |
Specification Test Results
Purity |
|
Appearance/Color | Solid. |
Solubility |
Soluble in DMSO. Slightly soluble ( |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...