PF-04217903 ≥95%

Supplier: ADIPOGEN CORP MS

Synonyms: 2-(4-(1-(Quinolin-6-ylmethyl)-1H-[1,2,3]triazolo[4,5-b]pyrazin-6-yl)-1H-pyrazol-1-yl)ethan-1-ol

SYN-1060-M001 SYN-1060-M005 SYN-1060-M010 SYN-1060-M050
102991-724EA 90 USD
102991-724 102991-726 102991-728 102991-730
PF-04217903 ≥95%
PF-04217903

PF04217903 is a novel orally bio-available, small molecule tyrosine kinase inhibitor with potential anti-neoplastic activity. It is an extremely selective MET inhibitor, with an IC(50) of 4.8nM.

Formula: C₁₉H₁₆N₈O
MW: 372.4 g/mol
Storage Temperature: Freezer
MDL Number: MFCD12407410
CAS Number: 956905-27-4

Order Now

Specification Test Results

Purity 95%
Appearance/Color Solid.
Solubility Soluble in DMSO. Slightly soluble (1mg/ml) in ethanol.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR