Merck-5 ≥95%
Supplier: ADIPOGEN CORP MS
Synonyms:
Pyridone 6, 2-(tert-Butyl)-9-fluoro-1,6-dihydro-7H-benzo[h]imidazo[4,5-f]isoquinolin-7-one, JAK Inhibitor
Merck 5, also known as Pyridone 6, is a potent and reversible ATP-competitive inhibitor of the JAK kinases (JAK1, JAK2, JAK3, and Tyk2) with IC(50) values in the low anomolar range. At higher values (IC(50)
Formula:
C₁₈H₁₆FN₃O MW: 309.3 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD17019334 CAS Number: 457081-03-7 |
Specification Test Results
Purity |
|
Appearance/Color | Solid. |
Solubility | Soluble in DMSO. |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...