Lapatinib ≥95%

Supplier: ADIPOGEN CORP MS
Warning

Synonyms: N-(3-Chloro-4-((3-fluorobenzyl)oxy)phenyl)-6-(5-(((2-(methylsulphonyl)ethyl)amino)methyl)furan-2-yl)quinazolin-4-amine, GW-572016

SYN-1052-M001 SYN-1052-M005 SYN-1052-M010 SYN-1052-M050
102991-654EA 74.77 USD
102991-654 102991-656 102991-658 102991-660
Lapatinib ≥95%
Lapatinib

Lapatinib is an ATP-competitive epidermal growth factor receptor (EGFR) and HER2/neu (ErbB-2) dual tyrosine kinase inhibitor, with IC(50) values of 10.8 and 9.2nM respectively. It is an orally active drug for treatment of breast cancer and other solid tumors.

Formula: C₂₉H₂₆ClFN₄O₄S
MW: 581.1 g/mol
Storage Temperature: Freezer
MDL Number: MFCD09264194
CAS Number: 231277-92-2

Order Now

Specification Test Results

Purity 95%
Appearance/Color Solid.
Solubility Soluble in DMSO or ethanol.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR