JNJ-38877605 ≥95%
Supplier: ADIPOGEN CORP MS
Synonyms:
6-(Difluoro(6-(1-methyl-1H-pyrazol-4-yl)-[1,2,4]triazolo[4,3-b]pyridazin-3-yl)methyl)quinoline
SYN-1047-M001
SYN-1047-M005
SYN-1047-M010
SYN-1047-M050
102991-614EA
134.31
USD
102991-614
102991-616
102991-618
102991-620
JNJ-38877605 ≥95%
JNJ-38877605
JNJ38877605 is an orally bio-available, small-molecule, c-MET tyrosine kinase inhibitor with potential anti-neoplastic activity. It is potent (c-MET enzyme and Phospho-MET IC(50) values of 4nM and 50nM, respectively) and highly specific (selective over 229 other kinases tested).
Formula:
C₁₉H₁₃F₂N₇ MW: 377.4 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD11977274 CAS Number: 943540-75-8 |
Specification Test Results
Purity |
|
Appearance/Color | Solid. |
Solubility |
Soluble in DMSO. Slightly soluble ( |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...