Dianilinopyrimidine_01 ≥95%

Supplier: ADIPOGEN CORP MS

Synonyms: CZC-8004, N2-(4-(Aminomethyl)phenyl)-5-fluoro-N4-phenylpyrimidine-2,4-diamine

SYN-1037-M001 SYN-1037-M005 SYN-1037-M010 SYN-1037-M050
102991-534EA 224.31 USD
102991-534 102991-536 102991-538 102991-540
Dianilinopyrimidine_01 ≥95%
Dianilinopyrimidine-01

Dianilinopyrimidine_01 is a pan tyrosine kinase inhibitor.

Formula: C₁₇H₁₆FN₅
MW: 309.3 g/mol
Storage Temperature: Freezer
MDL Number: MFCD16251195
CAS Number: 916603-07-1

Order Now

Specification Test Results

Purity 95%
Appearance/Color Solid.
Solubility Soluble in DMSO.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR