CYC116 ≥95%

Supplier: ADIPOGEN CORP MS

Synonyms: CYC-116, 4-Methyl-5-(2-((4-morpholinophenyl)amino)pyrimidin-4-yl)thiazol-2-amine

SYN-1034-M001 SYN-1034-M005 SYN-1034-M010 SYN-1034-M050
102991-510EA 90 USD
102991-510 102991-512 102991-514 102991-516
CYC116 ≥95%
CYC116

CYC116 is an orally active small molecule multi-kinase inhibitor with anti-neoplastic activity. CYC116 inhibits Aurora kinases A/B and vascular endothelial growth factor receptor 2 (VEGFR2), resulting in disruption of the cell cycle, rapid cell death, and the inhibition of angiogenesis.

Formula: C₁₈H₂₀N₆OS
MW: 368.5 g/mol
Storage Temperature: Freezer
MDL Number: MFCD14155805
CAS Number: 693228-63-6

Order Now

Specification Test Results

Purity 95%
Appearance/Color Solid.
Solubility Soluble in DMSO or ethanol.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR