CP-724714 ≥95%

Supplier: ADIPOGEN CORP MS

SYN-1033-M001 SYN-1033-M005 SYN-1033-M010 SYN-1033-M050
102991-502EA 119.08 USD
102991-502 102991-504 102991-506 102991-508
CP-724714 ≥95%
CP-724714

CP-724714 is an oral, selective and potent ErbB-2 (HER2) kinase inhibitor with an IC(50) of 10 nM. It has 640-fold selectivity for ErbB-2 (HER2) over EGFR, InsR, IRG-1R, PDGFR, VEGFR2, Abl, Src, c-Met.

Formula: C₂₇H₂₇N₅O₃
Storage Temperature: Freezer
MDL Number: MFCD11983048
CAS Number: 383432-38-0

Order Now

Specification Test Results

Purity 95%
Appearance/Color Solid.
Solubility Soluble in DMSO or ethanol.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR