Canertinib ≥95%

Supplier: ADIPOGEN CORP MS
Danger

Synonyms: N-(4-((3-Chloro-4-fluorophenyl)amino)-7-(3-morpholinopropoxy)quinazolin-6-yl)acrylamide

SYN-1030-M001 SYN-1030-M005 SYN-1030-M010 SYN-1030-M050
102991-478EA 74.77 USD
102991-478 102991-480 102991-482 102991-484
Canertinib ≥95%
Canertinib

Canertinib is an orally bio-available EGFR inhibitor with potential anti-neoplastic and radio-sensitizing activities. It has IC(50) values of 1.5nM and 9.0nM against EGFR and ErbB2 respectively, with no activity against PDGFR, FGFR, InsR, PKC, or CDK1/2/4.

Formula: C₂₄H₂₅ClFN₅O₃
MW: 486 g/mol
Storage Temperature: Freezer
MDL Number: MFCD09837878
CAS Number: 267243-28-7
UN: 3077
ADR: 9

Order Now

Specification Test Results

Purity >95%
Appearance/Color Solid.
Solubility Soluble in DMSO or ethanol.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR