BMS-3 ≥95%
Supplier: ADIPOGEN CORP MS
SYN-1023-M001
SYN-1023-M005
SYN-1023-M010
SYN-1023-M050
102991-422EA
119.08
USD
102991-422
102991-424
102991-426
102991-428
BMS-3 ≥95%
BMS-3
BMS-3 is a potent inhibitor of the LIM kinase. It has IC(50)'s of 5nM and 6nM for LIMK1 and LIMK2 respectively.
Formula:
C₁₇H₁₂Cl₂F₂N₄OS Storage Temperature: Freezer |
MDL Number:
MFCD17019326 CAS Number: 1338247-30-5 |
Specification Test Results
Purity |
|
Appearance/Color | Solid. |
Solubility | Soluble in DMSO. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...