Selumetinib ≥95%
Supplier: ADIPOGEN CORP MS
Selumetinib (AZD6244) is a second-generation, orally available, potent and selective ATP non-competitive inhibitor of MEK 1/2. In direct assays, AZD6244 selectively inhibits purified active MEK1 and MEK2 with an IC(50) of 14nM, and AZD6244 has reported IC(50) values of
Formula:
C₁₇H₁₅BrClFN₄O₃ Storage Temperature: Freezer |
MDL Number:
MFCD11977472 CAS Number: 606143-52-6 |
Specification Test Results
Purity |
|
Appearance/Color | Solid. |
Solubility |
Soluble in DMSO. Slightly soluble ( |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...