Tivozanib ≥95%
Supplier: ADIPOGEN CORP MS
Synonyms:
1-(2-Chloro-4-((6,7-dimethoxyquinolin-4-yl)oxy)phenyl)-3-(5-methylisoxazol-3-yl)urea, AV-951
SYN-1013-M001
SYN-1013-M005
SYN-1013-M010
SYN-1013-M050
102991-344EA
119.08
USD
102991-344
102991-346
102991-348
102991-350
Tivozanib ≥95%
Tivozanib
Tivozanib is a potent inhibitor of VEGFR-1, 2 & 3 (IC(50) 0.21, 0.16 and 0.24nM respectively).
Formula:
C₂₂H₁₉ClN₄O₅ MW: 454.9 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD15146788 CAS Number: 475108-18-0 |
Specification Test Results
Purity |
|
Appearance/Color | Solid. |
Solubility |
Soluble in DMSO. Slightly soluble ( |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...