AS-252424 ≥95%

Supplier: ADIPOGEN CORP MS
Warning

SYN-1009-M001 SYN-1009-M005 SYN-1009-M010 SYN-1009-M050
102991-314EA 96.92 USD
102991-314 102991-316 102991-318 102991-320
AS-252424 ≥95%
AS-252424

AS-252424 is a potent and selective PI3K inhibitor. It inhibits human recombinant PI3K alpha, beta, gamma and delta with IC(50) values of 30, 940, 20,000, and 20,000nM respectively.

Formula: C₁₄H₈FNO₄S
MW: 305.3 g/mol
Storage Temperature: Freezer
MDL Number: MFCD11045281
CAS Number: 900515-16-4

Order Now

Specification Test Results

Purity 95%
Appearance/Color Solid.
Solubility Soluble in DMSO. Slightly soluble (1mg/ml) in ethanol.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR