AMG-Tie2-1 ≥95%

Supplier: ADIPOGEN CORP MS

SYN-1008-M001 SYN-1008-M005 SYN-1008-M010
102991-308EA 292.15 USD
102991-308 102991-310 102991-312
AMG-Tie2-1 ≥95%
AMG-Tie2-1

AMG-Tie2-1 is a highly potent, non-selective inhibitor of Tie-2 (IC(50) 1nM).

Formula: C₂₅H₂₀F₃N₅O₂
MW: 479.5 g/mol
Storage Temperature: Freezer
CAS Number: 870223-96-4

Order Now

Specification Test Results

Purity 95%
Appearance/Color Solid.
Solubility Soluble in DMSO.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR