AG13958 ≥95%

Supplier: ADIPOGEN CORP MS

Synonyms: N-[2-fluoro-5-[[3-[(E)-2-pyridin-2-ylethenyl]-1H-indazol-6-yl]amino]phenyl]-2,5-dimethylpyrazole-3-carboxamide

SYN-1004-M001 SYN-1004-M005 SYN-1004-M010
102991-196EA 373.85 USD
102991-196 102991-198 102991-200
AG13958 ≥95%
AG13958

AG13958 is a VEGF inhibitor that was in clinical development for treatment of age-related macular degeneration (AMD). The drug displays low water solubility and so far in trials delivery has been difficult needing direct injection to be truly effective and it varied by the species used because of species specific differences in eye circulation and vascularization. As a VEGF inhibitor AG13958 was targeted to FLT/TYK receptor inhibition.

Formula: C₂₆H₂₂FN₇O
MW: 467.5 g/mol
Storage Temperature: Freezer
CAS Number: 319460-94-1

Order Now

Specification Test Results

Purity 95%
Appearance/Color Solid.
Solubility Soluble in DMSO.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR