Linifanib ≥95%
Supplier: ADIPOGEN CORP MS
Synonyms:
1-(4-(3-Amino-1H-indazol-4-yl)phenyl)-3-(2-fluoro-5-methylphenyl)urea, ABT-869
Linifanib (A 741439; A-741439; A741439; ABT-869; ABT869; RG3635) is an orally active multi-targeted receptor tyrosine kinase inhibitor for the treatment of various cancers. The compound is designed to inhibit vascular endothelial growth factor and platelet-derived growth factor receptors and is a multitargeted tyrosine kinase receptor inhibitor that suppresses FLT3 signaling. It is in phase III development for liver cancer and phase II development for non-small cell lung cancer, breast cancer, and colorectal cancer. Recent research also indicates that Linifanib inhibits proliferation and induces apoptosis in AML patient cells via reduction of AKT and GSK3beta phosphorylation and maybe influencial in AML therapy.
Formula:
C₂₁H₁₈FN₅O MW: 375.4 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD11840918 CAS Number: 796967-16-3 |
Specification Test Results
Purity |
|
Appearance/Color | Solid. |
Solubility | Soluble in DMSO or ethanol. |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...