Zinquin free acid ≥95% (by HPLC)

Supplier: ADIPOGEN CORP MS

Synonyms: Zinquin A

CDX-Z0014-M001 CDX-Z0014-M005 CDX-Z0014-M025
102991-260EA 113.54 USD
102991-260 102991-262 102991-264
Zinquin free acid ≥95% (by HPLC)
Zinquin

Used to detect zinc in cells by UV fluorescence.

Formula: C₁₉H₁₈N₂O₅S
MW: 386.42 g/mol
Storage Temperature: Freezer
MDL Number: MFCD03452711
CAS Number: 151606-29-0

Order Now

Specification Test Results

Source/Host Synthetic.
Purity >95% (HPLC)
Appearance/Color Solid.
Solubility Soluble in methanol or DMSO.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR