ZnAF-2, 6-Iso ≥95%
Supplier: ADIPOGEN CORP MS
Synonyms:
ZnAF-2, 6-{2-[Bis(2-pyridylmethyl)amino]ethylamino}fluorescein
CDX-Z0006-M001
CDX-Z0006-M005
CDX-Z0006-M025
102991-228EA
113.54
USD
102991-228
102991-230
102991-232
ZnAF-2, 6-Iso ≥95%
ZnAF-2, 6-Iso
A membrane-impermeable ethylenediaminofluoresceinyl compound that acts as a high-affinity Zn2+-specific fluorescent probe (Kd = 2.7 nM).
It shows low basal fluorescence and the fluorescence intensity increases ~51-fold upon stoichiometric (1:1) binding to Zn2+. Exhibits little affinity towards Ca2+, Mg2+, Na+, or K+. Spectral data: lambdaex 492nm; lambdaem 515nm in PBS.
Formula:
C₃₄H₂₈N₄O₅ Storage Temperature: Freezer |
MDL Number:
MFCD08277015 CAS Number: 321859-11-4 |
Specification Test Results
Source/Host | Synthetic. |
Purity |
|
Appearance/Color | Solid. |
Solubility | Soluble in DMSO. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...