ZnAF-1 ≥95% (by HPCE)
Supplier: ADIPOGEN CORP MS
Synonyms:
ZnAF-1, 5-Iso
CDX-Z0005-M005
CDX-Z0005-M125
102991-224EA
171.69
USD
102991-224
102991-226
ZnAF-1 ≥95% (by HPCE)
ZnAF-1
A membrane-impermeable ethylenediaminofluoresceinyl compound that acts as a high-affinity Zn2+-specific fluorescent probe. Spectral data: lambdaex 492 nm; lambdaem 514 nm in PBS.
Formula:
C₃₄H₂₈N₄O₅ MW: 572.61 g/mol Storage Temperature: Freezer |
CAS Number:
321859-09-0 |
Specification Test Results
Source/Host | Synthetic. |
Purity |
|
Appearance/Color | Solid. |
Solubility | Soluble in DMSO. |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...