Tirofiban ≥99% (by HPLC)

Supplier: ADIPOGEN CORP MS
Warning

Synonyms: (S)-2-(Butylsulphonamido)-3-(4-(4-(piperidin-4-yl)butoxy)phenyl)propanoic acid, MK-383, L-7462

CDX-T0153-M005 CDX-T0153-M025 CDX-T0153-M100
102991-154EA 139.85 USD
102991-154 102991-156 102991-158
Tirofiban ≥99% (by HPLC)
Tirofiban

Synthetic non-peptide selective glycoprotein (GP) IIb/IIIa receptor antagonist/inhibitor. Anticoagulant, specifically, an anti-platelet aggregation inhibitor.

Formula: C₂₂H₃₆N₂O₅S
MW: 440.60 g/mol
Storage Temperature: Freezer
MDL Number: MFCD05237246
CAS Number: 144494-65-5

Order Now

Specification Test Results

Source/Host Synthetic
Purity >99% (HPLC)
Appearance/Color Off-white powder.
Solubility Sligthly soluble in ethanol or water.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR