2-(Tosyloxy)cyclohexanone ≥98%

Supplier: ADIPOGEN CORP MS
Warning

Synonyms: CTK3E4439

CDX-T0131-G001 CDX-T0131-G005
102991-134EA 139.85 USD
102991-134 102991-136
2-(Tosyloxy)cyclohexanone ≥98%
2-(Tosyloxy)cyclohexanone

Alpha-​tosyloxy ketone used for synthesis.

Formula: C₁₃H₁₆O₄S
Storage Temperature: Freezer
CAS Number: 81447-34-9

Order Now

Specification Test Results

Source/Host Synthetic.
Purity 98% (HPLC)
Appearance/Color White powder.
Solubility Soluble in chloroform.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR